missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Homopiperazinylmethyl)indole, 95%, Thermo Scientific™

Catalog Number 15459608
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1183967-11-4
3-(1,4-Diazepan-1-ylmethyl)indole
This item is not returnable. View return policy
1183967-11-4
3-(1,4-Diazepan-1-ylmethyl)indole

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Beilstein 11950916
Formula Weight 229.32
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Homopiperazinylmethyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'