missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,5,6-tetrahydro-4-pyridyl)-6-nitroindole, 97%

Brand:  Alfa Aesar™ H63240.06

Code : NEW

Additional Details : CAS Number : 1342806-94-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428186

  • £478.06 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,5,6-tetrahydro-4-pyridyl)-6-nitroindole
343.38
1-Boc-4-(6-nitro-3-indolyl)-1,2,3,6-tetrahydropyridine
1342806-94-3
MFCD28098701

Specifications

3-(1-Boc-1,2,5,6-tetrahydro-4-pyridyl)-6-nitroindole
Solid
343.38
1-Boc-4-(6-nitro-3-indolyl)-1,2,3,6-tetrahydropyridine
1342806-94-3
97%
MFCD28098701
5g
Videos
SDS