missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole, 97%

Brand:  Alfa Aesar™ H63242.03

Code : NEW

Additional Details : CAS Number : 935679-13-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448186

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole
316.37
15719293
935679-13-3
MFCD28098700
1-Boc-4-(6-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole
Solid
316.37
15719293
1g
935679-13-3
97%
MFCD28098700
1-Boc-4-(6-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine
Videos
SDS