missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole, 97%

Brand:  Alfa Aesar™ H63242.MD

Code : NEW

Additional Details : CAS Number : 935679-13-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15468186

  • £14.95 / 250mg


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole
316.37
15719293
935679-13-3
MFCD28098700
1-Boc-4-(6-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-fluoroindole
Solid
316.37
15719293
250mg
935679-13-3
97%
MFCD28098700
1-Boc-4-(6-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine
Videos
SDS