missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole, 95%

Brand:  Alfa Aesar™ H63306.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15468226

  • £59.78 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole
MFCD28098702
1-Boc-4-(6-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-6-carbonitrile
323.39
UN3439

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole
95%
MFCD28098702
Off-white to pale yellow
1g
Solid
323.39
UN3439
1-Boc-4-(6-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-6-carbonitrile
Videos
SDS