missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole, 97%

Brand:  Alfa Aesar™ H63145.06

Code : NEW

Additional Details : CAS Number : 345235-82-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458146

  • £298.92 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole
377.28
1-Boc-4-(6-bromo-3-indolyl)-1,2,3,6-tetrahydropyridine
345235-82-7
MFCD28100487

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole
Solid
377.28
1-Boc-4-(6-bromo-3-indolyl)-1,2,3,6-tetrahydropyridine
345235-82-7
97%
MFCD28100487
5g
Videos
SDS