missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63636.06

Code : NEW

Additional Details : CAS Number : 151273-41-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15498416

  • £298.92 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole
343.38
9940902
151273-41-5
MFCD26940339
1-Boc-4-(5-nitro-3-indolyl)-1,2,3,6-tetrahydropyridine

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole
Solid
343.38
9940902
5g
151273-41-5
97%
MFCD26940339
1-Boc-4-(5-nitro-3-indolyl)-1,2,3,6-tetrahydropyridine
Videos
SDS