missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-fluoroindole, 97%

Brand:  Alfa Aesar™ H63153.MD

Code : NEW

Additional Details : CAS Number : 935678-62-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15428156

  • £14.95 / 250mg


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-fluoroindole
316.37
9936989
935678-62-9
MFCD24468951
1-Boc-4-(5-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-fluoroindole
Solid
316.37
9936989
250mg
935678-62-9
97%
MFCD24468951
1-Boc-4-(5-fluoro-3-indolyl)-1,2,3,6-tetrahydropyridine
Videos
SDS