missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole, 95%

Brand:  Alfa Aesar™ H63810.03

Code : NEW

Additional Details : CAS Number : 345235-76-9 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15408536

  • £59.78 / 1g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
323.39
UN3439
1-Boc-4-(5-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile
345235-76-9
MFCD24467119
11211398

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
Solid
323.39
UN3439
Off-white to yellow
1g
345235-76-9
95%
MFCD24467119
11211398
1-Boc-4-(5-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile
Videos
SDS