Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63810.06

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 345235-76-9 Weight: 0.00500kg


Quantity 5g

Product Code. 15418536

Quantity Price
1 £ 298.92 / 5g
Estimated Shipment date
from Supplier 06-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
CAS 345235-76-9
Formula Weight 323.39
MDL Number MFCD24467119
UN Number UN3439
Beilstein 11211398
Synonym 1-Boc-4-(5-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile

Specification

Formula Weight 323.39
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Chemical Name or Material 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
Beilstein 11211398
Health Hazard 1 Warning
MDL Number MFCD24467119
Quantity 5g
UN Number UN3439
Color Off-white to yellow
Physical Form Solid
Synonym 1-Boc-4-(5-cyano-3-indolyl)-1,2,3,6-tetrahydropyridine; 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile
CAS 345235-76-9
Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.