missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-bromoindole, 97%

Brand:  Alfa Aesar™ H63327.MD

Code : NEW

Additional Details : CAS Number : 886361-90-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15468246

  • £14.95 / 250mg


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-bromoindole
377.28
9936283
886361-90-6
MFCD08276937
1-Boc-4-(5-bromo-3-indolyl)-1,2,3,6-tetrahydropyridine

Specifications

3-(1-Boc-1,2,3,6-tetrahydro-4-pyridyl)-5-bromoindole
Solid
377.28
9936283
250mg
886361-90-6
97%
MFCD08276937
1-Boc-4-(5-bromo-3-indolyl)-1,2,3,6-tetrahydropyridine
Videos
SDS