missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole, 97%

Brand:  Alfa Aesar™ H63375.MD

Code : NEW

Additional Details : CAS Number : 181184-09-8 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15478276

  • £14.95 / 250mg


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole
313.4
UN3439
181184-09-8
MFCD28126771
3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole-6-carbonitrile

Specifications

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-cyanoindole
Solid
313.4
UN3439
250mg
181184-09-8
97%
MFCD28126771
Off-white to yellow
3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole-6-carbonitrile
Videos
SDS