missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole, 97%

Brand:  Alfa Aesar™ H63696.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428456

  • £298.92 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole
367.28
MFCD28098694
6-Bromo-3-(1-benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole

Specifications

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-6-bromoindole
97%
MFCD28098694
6-Bromo-3-(1-benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole
Solid
367.28
5g
Videos
SDS