missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole, 95%

Brand:  Alfa Aesar™ H63995.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15448626

  • £59.78 / 1g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
MFCD28098696
13255692
313.4
UN3439
3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile

Specifications

3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)-5-cyanoindole
95%
MFCD28098696
13255692
1g
Solid
313.4
UN3439
Off-white to orange
3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)indole-5-carbonitrile
Videos
SDS