missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63266.03

Code : NEW

Additional Details : CAS Number : 915037-53-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468206

  • £59.78 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole
915037-53-5
300.31
MFCD28126767

Specifications

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole
915037-53-5
97%
MFCD28126767
1g
Solid
300.31
Videos
SDS