missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-[1-(4,4-Dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino]-Fmoc-L-alanine, 95%

Brand:  Alfa Aesar™ H63926.MD

Code : NEW

Additional Details : CAS Number : 607366-20-1 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15455645

  • £26.71 / 250mg


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

3-[1-(4,4-Dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino]-Fmoc-L-alanine
532.63
Fmoc-Dap(ivDde)-OH; (S)-3-[(1-[4,4-Dimethyl-2,6-dioxocyclohexylidene]-3-methylbutyl)amino]-2-(Fmoc-amino)propionic acid
607366-20-1
MFCD01631659

Specifications

3-[1-(4,4-Dimethyl-2,6-dioxocyclohexylidene)-3-methylbutylamino]-Fmoc-L-alanine
Solid
532.63
Fmoc-Dap(ivDde)-OH; (S)-3-[(1-[4,4-Dimethyl-2,6-dioxocyclohexylidene]-3-methylbutyl)amino]-2-(Fmoc-amino)propionic acid
607366-20-1
95%
MFCD01631659
250mg
Videos
SDS