missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Oxazol-5-yl)benzoic acid, ≥97%, Maybridge™

£52.00 - £157.00

Chemical Identifiers

CAS 252928-82-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD06797481
InChI Key GDGXRJDVOKNSCX-UHFFFAOYSA-N
Synonym 3-1,3-oxazol-5-yl benzoic acid, 3-oxazol-5-yl benzoic acid, 3-5-oxazolyl benzoic acid, benzoic acid, 3-5-oxazolyl, 3-5-oxazolyl benzoicacid, benzoicacid, 3-5-oxazolyl
PubChem CID 7127813
IUPAC Name 3-(1,3-oxazol-5-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CN=CO2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10094562
SDS View Documents
Maybridge
CC35801CB
250mg Amber Glass Bottle
£52.00
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-10-2021
Please to purchase this item. Need a web account? Register with us today!
10107852
SDS View Documents
Maybridge
CC35801DA
1g Amber Glass Bottle
£157.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 20-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

252928-82-8
189.17
GDGXRJDVOKNSCX-UHFFFAOYSA-N
7127813
C1=CC(=CC(=C1)C(=O)O)C2=CN=CO2
C10H7NO3
MFCD06797481
3-1,3-oxazol-5-yl benzoic acid, 3-oxazol-5-yl benzoic acid, 3-5-oxazolyl benzoic acid, benzoic acid, 3-5-oxazolyl, 3-5-oxazolyl benzoicacid, benzoicacid, 3-5-oxazolyl
3-(1,3-oxazol-5-yl)benzoic acid

Specifications

3-(1, 3-Oxazol-5-yl)benzoic acid
250mg
96.5
97%
MFCD06797481
GDGXRJDVOKNSCX-UHFFFAOYSA-N
3-(1,3-oxazol-5-yl)benzoic acid
7127813
97%
Amber Glass Bottle
252928-82-8
100.0
C10H7NO3
3-1,3-oxazol-5-yl benzoic acid, 3-oxazol-5-yl benzoic acid, 3-5-oxazolyl benzoic acid, benzoic acid, 3-5-oxazolyl, 3-5-oxazolyl benzoicacid, benzoicacid, 3-5-oxazolyl
C1=CC(=CC(=C1)C(=O)O)C2=CN=CO2
189.17
189.17
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 3-(1,3-Oxazol-5-yl)benzoic acid

SDS
Product Certifications