missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)thiophene-2-sulfonyl chloride, ≥97%, Thermo Scientific™

Catalog Number 10051574
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
103011-38-7
C7H7ClO4S2
254.70
MFCD08271969
RRTOURREASJQSR-UHFFFAOYSA-N
3-1,3-dioxolan-2-yl thiophene-2-sulfonyl chloride, chloro 3-1,3-dioxolan-2-yl 2-thienyl sulfone, 3-1,3-dioxolan-2-yl-thiophene-2-sulfonyl chloride
10538915
3-(1,3-dioxolan-2-yl)thiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=C(C=CS1)C1OCCO1
This item is not returnable. View return policy
103011-38-7
C7H7ClO4S2
254.70
MFCD08271969
RRTOURREASJQSR-UHFFFAOYSA-N
3-1,3-dioxolan-2-yl thiophene-2-sulfonyl chloride, chloro 3-1,3-dioxolan-2-yl 2-thienyl sulfone, 3-1,3-dioxolan-2-yl-thiophene-2-sulfonyl chloride
10538915
3-(1,3-dioxolan-2-yl)thiophene-2-sulfonyl chloride
ClS(=O)(=O)C1=C(C=CS1)C1OCCO1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 254.71
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Dioxolan-2-yl)thiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'