missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3541034
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
934570-44-2
C8H8O4S
200.21
MFCD09702391
ZODQJPSGAHBVHU-UHFFFAOYSA-N
3-1,3-dioxolan-2-yl thiophene-2-carboxylic acid, 3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid, 2-2-carboxythien-3-yl-1,3-dioxolane, 3-1,3-dioxolan-2-yl thiophene-2-carboxylicacid, 2-thiophenecarboxylicacid, 3-1,3-dioxolan-2-yl
24229593
3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
OC(=O)C1=C(C=CS1)C1OCCO1
This item is not returnable. View return policy
934570-44-2
C8H8O4S
200.21
MFCD09702391
ZODQJPSGAHBVHU-UHFFFAOYSA-N
3-1,3-dioxolan-2-yl thiophene-2-carboxylic acid, 3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid, 2-2-carboxythien-3-yl-1,3-dioxolane, 3-1,3-dioxolan-2-yl thiophene-2-carboxylicacid, 2-thiophenecarboxylicacid, 3-1,3-dioxolan-2-yl
24229593
3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
OC(=O)C1=C(C=CS1)C1OCCO1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.21
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Dioxolan-2-yl)-2-thiophenecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'