missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1,3-Benzothiazol-2-yl)aniline, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 226.3 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1, 3-benzothiazol-2-yl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'