missing translation for 'onlineSavingsMsg'
Learn More

3-(1,2,3,6-Tetrahydro-4-pyridyl)indole, 95%, Thermo Scientific™

Catalog Number 15400729
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
65347-55-9
C13H14N2
198.269
MFCD03695471
CIRSPTXGPFAXRE-UHFFFAOYSA-N
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
2761023
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
C1CNCC=C1C2=CNC3=CC=CC=C32
This item is not returnable. View return policy
65347-55-9
C13H14N2
198.269
MFCD03695471
CIRSPTXGPFAXRE-UHFFFAOYSA-N
3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, chembl27811, 3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 3-4-1,2,5,6-tetrahydropyridyl indole, thpindole, d02mvr, 3-1,2,3,6-tetrahydropyridin-4-yl indole, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indol, 3-1,2,3,6-tetrahydro-4-pyridinyl 1h-indole
2761023
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
C1CNCC=C1C2=CNC3=CC=CC=C32

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 958099
Formula Weight 198.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1,2,3,6-Tetrahydro-4-pyridyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'