missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2xYT Broth (Powder/Molecular Genetics), Fisher BioReagents
For use in molecular genetics
£23.08 - £1158.16
Specifications
Content And Storage | <4°C |
---|---|
Additive | 2xyt Broth |
Form | Powder |
Formulation | 31g/L |
For Use With (Application) | Molecular Genetics |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
10123103
|
Fisher Bioreagents
BP2467-10 |
Poly Pail | 10,000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10346793
|
Fisher Bioreagents
BP2467-100 |
Poly Bottle | 100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10011223
|
Fisher Bioreagents
BP2467-2 |
Poly Bottle | 2000 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10617633
|
Fisher Bioreagents
BP2467-500 |
Poly Bottle | 500 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Formulation Usage: 31.0g/L
Specifications
<4°C | |
Powder | |
Molecular Genetics |
2xyt Broth | |
31g/L |