missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2xYT Agar (Powder/Molecular Genetics), Fisher BioReagents
£21.36 - £242.96
Specifications
Additive | 2xyt Agar |
---|---|
Form | Powder |
For Use With (Application) | Molecular Genetics |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
11403064
|
Fisher Bioreagents
BP2466-2 |
2000 g | Poly Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10724534
|
Fisher Bioreagents
BP2466-100 |
100 g | Poly Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10041793
|
Fisher Bioreagents
BP2466-500 |
500 g | Poly Bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
2xyt Agar | |
Powder | |
Molecular Genetics |