(2S,5S)-(+)-Hexanediol, 99%, 99% e.e., ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 318760010

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 34338-96-0 Weight: 0.05100kg


Packaging Glass bottle
Quantity 1g

Product Code. 10731555

Quantity Price
1 £ 83.7 / 1g
EU Stock 4
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (2S, 5S)-(+)-Hexanediol
Name Note 99% (99% E.E.)
CAS 34338-96-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Formula Weight 118.18
MDL Number MFCD00082583

Specification

Assay ≥98.5%
Physical Form Crystals
Formula Weight 118.18
Quantity 1g
Color White
Specific Rotation Condition + 36.00 (20.00°C c=5,CHCl3)
CAS Min % 98.5
Specific Rotation +35° to +37° (20°C, 589nm) (c=5, CHCl3)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Infrared Spectrum Authentic
CAS 34338-96-0
MDL Number MFCD00082583
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Glass bottle
Molecular Formula C6H14O2
CAS Max % 100.0
Health Hazard 1 HYGROSCOPIC
Name Note 99% (99% E.E.)
Enantiomeric Excess 98.5% min. (GC)
Chemical Name or Material (2S, 5S)-(+)-Hexanediol
TSCA TSCA
Melting Point 50° to 53°C
Documentation

Technical documents

Safety and Handling

HYGROSCOPIC

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Special Offers