missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane, 97%, Thermo Scientific™

Catalog Number 11905961
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
695816-47-8
C15H21O2P
264.3
JAKXTLMNIHPLEU-RYUDHWBXSA-N
2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
16218307
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
This item is not returnable. View return policy
695816-47-8
C15H21O2P
264.3
JAKXTLMNIHPLEU-RYUDHWBXSA-N
2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, s,s-me-rajphos, 2-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2s,5s-+-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, kanata purity, 1-2-1,3-dioxolane-2-yl phenyl-2beta,5alpha-dimethyltetrahydro-1h-phosphole, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
16218307
2-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1030g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 mg
Specific Gravity 1.103
Formula Weight 264.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-dimethylphospholane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'