missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%

Brand:  Alfa Aesar™ H63462.ME

Code : NEW

Additional Details : CAS Number : 695816-47-8 Weight : 0.00050kg

Quantity 500mg

 View more versions of this product

Product Code. 15488316

  • £171.30 / 500mg


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
264.3
S,S-Me-RajPhos
695816-47-8
MFCD09265097

Specifications

(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
500mg
1.103g/mL
97%
MFCD09265097
1.5715
Air Sensitive
695816-47-8
Liquid
264.3
S,S-Me-RajPhos
Pale yellow to colorless
Videos
SDS