missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 97%, Thermo Scientific™

Catalog Number 11915941
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
1314246-02-0
C17H25O2P
292.36
MFCD09265099
UKBFKBMQNUOKTD-UHFFFAOYNA-N
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
16218309
1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-diethylphospholane
CCC1CCC(CC)P1C1=CC=CC=C1C1OCCO1
This item is not returnable. View return policy
1314246-02-0
C17H25O2P
292.36
MFCD09265099
UKBFKBMQNUOKTD-UHFFFAOYNA-N
2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
16218309
1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-diethylphospholane
CCC1CCC(CC)P1C1=CC=CC=C1C1OCCO1
Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 292.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'