missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt, 95%, 98% ee, ACROS Organics™

Brand:  Acros Organics 362012500

Code : 29

Additional Details : CAS Number : 441044-14-0 Weight : 0.05025kg

Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10354452

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
95%
352.43
95%
C11H19NO5·C7H9N

Specifications

(2S, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
97.5% (HPLC) min.
Glass bottle
95%
C11H19NO5·C7H9N
250mg
441044-14-0
Authentic
Crystalline Powder
95%
352.43
White
Videos
SDS