missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-4510416
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
104195-80-4
C5H11NO3
133.15
FYCWLJLGIAUCCL-IMJSIDKUSA-N
2s,3s-2-amino-3-methoxybutanoic acid, 2s,3s-2-amino-3-methoxy-butanoic acid, allo-o-methyl-l-thr, l-allo-o-methylthreonine, 2s,3s-2-amino-3-methoxybutanoic acid
151207
(2S,3S)-2-amino-3-methoxybutanoic acid
CC(C(C(=O)O)N)OC
This item is not returnable. View return policy
104195-80-4
C5H11NO3
133.15
FYCWLJLGIAUCCL-IMJSIDKUSA-N
2s,3s-2-amino-3-methoxybutanoic acid, 2s,3s-2-amino-3-methoxy-butanoic acid, allo-o-methyl-l-thr, l-allo-o-methylthreonine, 2s,3s-2-amino-3-methoxybutanoic acid
151207
(2S,3S)-2-amino-3-methoxybutanoic acid
CC(C(C(=O)O)N)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 220.0°C to 224.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH(NH2)COOH
Quantity 250 mg
Specific Rotation Condition + 27.50 (25.00°C c=0.6,6N HCl)
Specific Rotation + 27.50
Solubility Information Solubility in water: soluble in water.
Formula Weight 133.15
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material (2S, 3S)-2-Amino-3-methoxybutanoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'