missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(+)-2,3-Butanediol, 99%, Thermo Scientific Chemicals

Catalog Number p-77901
Click to view available options
Quantity:
500 mg
2.5 g
This item is not returnable. View return policy
19132-06-0
C4H10O2
90.12
MFCD00063648
OWBTYPJTUOEWEK-IMJSIDKUSA-N
2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol
439888
CHEBI:16812
(2S,3S)-butane-2,3-diol
CC(C(C)O)O
This item is not returnable. View return policy
19132-06-0
C4H10O2
90.12
MFCD00063648
OWBTYPJTUOEWEK-IMJSIDKUSA-N
2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol
439888
CHEBI:16812
(2S,3S)-butane-2,3-diol
CC(C(C)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 179.0°C to 182.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH(OH)CH3
Refractive Index 1.4309 to 1.4329
Quantity 500 mg
Beilstein 01, 2183
Fieser 01,82
Merck Index 15, 1571
Specific Gravity 0.98
Specific Rotation + 13.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material (2S, 3S)-(+)-2, 3-Butanediol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'