missing translation for 'onlineSavingsMsg'
Learn More

(2R)-(-)-Glycidyl tosylate, 99+%, ACROS Organics™

Brand:  Acros Organics 264590010

Code : 29

Additional Details : CAS Number : 113826-06-5 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10043220

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(2R)-(-)-Glycidyl tosylate
99.0
≥99%
C10H12O4S
MFCD00010834
113826-06-5
100.0
99+%
228.27
(S)-Oxiranemethanol 4-methylbenzenesulfonate

Specifications

(2R)-(-)-Glycidyl tosylate
99.0
Authentic
≥99%
Glass bottle
228.27
Solubility in water: decomposes. Other solubilities: soluble in chloroform
− 18.00
White
1g
113826-06-5
100.0
Crystalline Powder
99+%
C10H12O4S
MFCD00010834
(S)-Oxiranemethanol 4-methylbenzenesulfonate
− 18.00 (20.00°C c=2.5,CHCl3)
45.0° to 48.0°C
>110°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (2R)-(-)-Glycidyl tosylate

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS