missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Bornane-10,2-sultam, 99+%, Thermo Scientific™

Catalog Number p-3756974
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
94594-90-8
MFCD00066271
DPJYJNYYDJOJNO-SZBHIRRCSA-N
5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, --10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
54579826
CC1(C2CCC13CS(=O)(=O)NC3C2)C
This item is not returnable. View return policy
94594-90-8
MFCD00066271
DPJYJNYYDJOJNO-SZBHIRRCSA-N
5r,7r-10,10-dimethyl-3??-thia-4-azatricyclo 5.2.1.0 1 ,? decane-3,3-dione, --10,2-camphorsultam;--exo-10,2-bornanesultam; 3as-3a, a,6, a,7a, a-hexahydro-8,8-dimethyl-3h-3a,6-methano-2,1-benzisothiazole-2,2-dioxide; 1s,5r-10,10-dimethyl-3-thia-4-azatricyclo 5.2.1.01,5 decane 3,3-dioxide
54579826
CC1(C2CCC13CS(=O)(=O)NC3C2)C
Melting Point 181°C to 185°C
Color Colorless
Enantiomeric Excess > 99%
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C10H17NO2S
Quantity 1 g
Specific Rotation -33
Molecular Weight (g/mol) 215.32
Formula Weight 215.32
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (2R)-Bornane-10, 2-sultam, ≥99% ee
Show More Show Less
compliance-icons
Product Identifier
  • (2R)-Bornane-10,2-sultam
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'