missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane, 97%

Brand:  Alfa Aesar™ H63024.MC

Code : NEW

Additional Details : CAS Number : 1217655-83-8 Weight : 0.00010kg

Quantity 100mg

 View more versions of this product

Product Code. 15488096

  • £24.49 / 100mg


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane
292.35
R,R-Et-RajPhos
1217655-83-8
MFCD09265098

Specifications

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane
1.5620
97%
292.35
Pale yellow to colorless
100mg
1217655-83-8
Liquid
1.073g/mL
MFCD09265098
R,R-Et-RajPhos
Air Sensitive
Videos
SDS