missing translation for 'onlineSavingsMsg'
Learn More

(2R,4R)-n-boc-4-hydroxypiperidine-2-carboxylic Acid Benzylamine Salt, 95%, ACROS Organics™

Brand:  Acros Organics 380762500

Code : 29

Additional Details : CAS Number : 441044-16-2 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 1 Pack group : III


Quantity 250mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10197582

  • £97.20 / 250mg


Estimated Shipment
18-05-2018
Add to basket


Specifications

Chemical Identifiers

(2R, 4R)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
94.0
≥94%
C11H19NO5·C7H9N
MFCD02259745
441044-16-2
100.0
95%
352.43

Specifications

(2R, 4R)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid benzylamine salt
94.0
97.5% min. (HPLC)
Glass bottle
≥94%
C11H19NO5·C7H9N
MFCD02259745
White
441044-16-2
100.0
Authentic
Crystalline Powder
95%
352.43
250mg
Videos
SDS