Learn More
(2R,4R)-APDC, Tocris Bioscience™
Highly selective group II agonist
Brand:  Tocris (Bio-Techne) 1208/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13448873
  • £238.14 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
A highly selective and relatively potent group II metabotropic glutamate receptor agonist. EC50 values are 0.4, 0.4, > 100, > 100, > 300 and > 300 μM for human mGlu2, mGlu3, mGlu1, mGlu5, mGlu4 and mGlu7 receptors respectively. Centrally active following systemic administration in vivo. Also available as part of the Group II mGlu Receptor Tocriset™ .
Specifications
Chemical Identifiers
169209-63-6
174.156
2r,4r-apdc, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylic acid, chembl40086, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylate, 2r, 4r-apdc monohydrate, 4-amino-2,4-pyrrolidinedicarboxylic acid, d06tod, d0ek6p, 2r, 4r-apdc
CHEBI:40139
C1C(NCC1(C(=O)O)N)C(=O)O
C6H10N2O4
XZFMJVJDSYRWDQ-AWFVSMACSA-N
5310984
(2R,4R)-4-aminopyrrolidine-2,4-dicarboxylic acid
Specifications
10mg
169209-63-6
2r,4r-apdc, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylic acid, chembl40086, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylate, 2r, 4r-apdc monohydrate, 4-amino-2,4-pyrrolidinedicarboxylic acid, d06tod, d0ek6p, 2r, 4r-apdc
C1C(NCC1(C(=O)O)N)C(=O)O
174.156
CHEBI:40139
>98%
(2R, 4R)-APDC
C6H10N2O4
XZFMJVJDSYRWDQ-AWFVSMACSA-N
(2R,4R)-4-aminopyrrolidine-2,4-dicarboxylic acid
5310984
174.16