missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, 85%, Tech., Thermo Scientific™

Catalog Number p-2689191
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
205639-93-6
MFCD00142975
BBLPZPQHEWQLGM-VZXYPILPSA-M
potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
23666349
potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]
This item is not returnable. View return policy
205639-93-6
MFCD00142975
BBLPZPQHEWQLGM-VZXYPILPSA-M
potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
23666349
potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 80.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H17KO4
Quantity 250 mg
Molecular Weight (g/mol) 264.35
Formula Weight 264.35
Percent Purity 85%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, tech., 85%
Show More Show Less
compliance-icons
Product Identifier
  • (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene, potassium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'