missing translation for 'onlineSavingsMsg'
Learn More

2H-Pyran-2-one, 97%, Thermo Scientific Chemicals

Catalog Number p-3523303
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
504-31-4
C5H4O2
96.09
MFCD00006639
ZPSJGADGUYYRKE-UHFFFAOYSA-N
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
68154
CHEBI:37965
pyran-2-one
C1=CC(=O)OC=C1
This item is not returnable. View return policy
504-31-4
C5H4O2
96.09
MFCD00006639
ZPSJGADGUYYRKE-UHFFFAOYSA-N
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
68154
CHEBI:37965
pyran-2-one
C1=CC(=O)OC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 8°C to 9°C
Color Yellow to Brown
Density 1.2100g/mL
Boiling Point 104°C (30.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.531
Quantity 1 g
Specific Gravity 1.21
Formula Weight 96.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2H-Pyran-2-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'