missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2H)1,4-Benzothiazin-3(4H)-one, 99%

Brand:  Alfa Aesar™ A11091.14

Code : NEW

Additional Details : CAS Number : 5325-20-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11418963

  • £50.99 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

(2H)1,4-Benzothiazin-3 (4H)-one
C8H7NOS
MFCD00006698
5325-20-2
165.22
131642

Specifications

(2H)1,4-Benzothiazin-3 (4H)-one
25g
99%
165.22
131642
176° to 178°C
5325-20-2
C8H7NOS
MFCD00006698
Videos
SDS