missing translation for 'onlineSavingsMsg'
Learn More

(2E,5E)-2,5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone, 95%, Thermo Scientific™

Catalog Number p-3211002
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261360-66-1
C27H34N2O
402.58
MFCD06658953
KKLBPVXKMBLCQX-BLVCXSLXSA-N
cyclopentanone, 2,5-bis 4-diethylamino phenyl methylene, 2e,5e-2,5-bis 4-diethylamino phenyl methylene cyclopentanone, 2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-bis 4-diethylaminobenzylidene cyclopentanone 17, 2,5-bis e-4-diethylamino benzylidene cyclopentanone, 2e 5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2e,5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-di e-1-4-diethylaminophenyl methylidene-1-cyclopentanone, 2e,5e-2,5-bis 4-diethylamino phenyl methylidene cyclopentan-1-one
5702661
(2E,5E)-2,5-bis[[4-(diethylamino)phenyl]methylidene]cyclopentan-1-one
CCN(CC)C1=CC=C(C=C1)C=C2CCC(=CC3=CC=C(C=C3)N(CC)CC)C2=O
This item is not returnable. View return policy
261360-66-1
C27H34N2O
402.58
MFCD06658953
KKLBPVXKMBLCQX-BLVCXSLXSA-N
cyclopentanone, 2,5-bis 4-diethylamino phenyl methylene, 2e,5e-2,5-bis 4-diethylamino phenyl methylene cyclopentanone, 2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-bis 4-diethylaminobenzylidene cyclopentanone 17, 2,5-bis e-4-diethylamino benzylidene cyclopentanone, 2e 5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2e,5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-di e-1-4-diethylaminophenyl methylidene-1-cyclopentanone, 2e,5e-2,5-bis 4-diethylamino phenyl methylidene cyclopentan-1-one
5702661
(2E,5E)-2,5-bis[[4-(diethylamino)phenyl]methylidene]cyclopentan-1-one
CCN(CC)C1=CC=C(C=C1)C=C2CCC(=CC3=CC=C(C=C3)N(CC)CC)C2=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 186°C to 189°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 402.58
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (2E, 5E)-2, 5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone
Show More Show Less
compliance-icons
Product Identifier
  • (2E,5E)-2,5-Bis¢(4-(diethylamino)phenyl)-methylene!cyclopentanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'