missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
£15.58 - £2282.35
Specifications
Content And Storage | Store at room temperature. |
---|---|
Sterilization Method | Irradiated |
Film Description | ASI 26 |
Product Code | Brand | Capacity (Metric) | Tubing Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Tubing Style | Price | Quantity & Availability | ||||
17860787
|
Thermo Scientific™
B100051-I |
560 L | No Tubing |
£481.00
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17830787
|
Thermo Scientific™
B100049-I |
340 L | No Tubing |
£2282.35
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17880777
|
Thermo Scientific™
B100038-I |
130 L | No Tubing |
£300.80
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17840787
|
Thermo Scientific™
B100050-I |
400 L | No Tubing |
£372.20
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17860777
|
Thermo Scientific™
B100037-I |
200 L | No Tubing |
£15.58
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17810787
|
Thermo Scientific™
B100048-I |
50 L | No Tubing |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Store at room temperature. | |
Irradiated | |
ASI 26 |