missing translation for 'onlineSavingsMsg'
Learn More
Learn More
28 Elements Solution, 100mg/L HNO3 matrix, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Concentration | 100mg/L |
Quantity | 100 mL |
Solution Contains | Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb Sb Se Sr, Ti, Tl, V, Zn for ICP 100mg/L HNO3 matrix |
Safety and Handling
Product Identifier
- Multi-element Standard Solution for ICP. 28 Components in HNO3 5% (MS 102)
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'