missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 20mm Crimp Cap

Aluminum crimp closures provide a secure leak-resistant seal.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12616765

  • £43.35 / Pack of 1000


Estimated Shipment
04-06-2021
Add to basket

Description

Description

  • 20mm
  • Aluminium Cap
  • Aluminum seals must be applied with a crimping tool
Specifications

Specifications

Gold Lacquered
Aluminium
100
20 mm
Crimp Cap
ND20
Videos
SDS
Documents