missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 20mm Crimp Cap
Aluminum crimp closures provide a secure leak-resistant seal.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- 20mm
- Aluminium Cap
- Aluminum seals must be applied with a crimping tool
Specifications
Green Lacquered | |
Aluminium | |
100 |
20 mm | |
Crimp Cap | |
ND20 |