missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Pore Size: Under 10mm

Brand:  RETSCH 60.149.005000

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 10630272

  • £135.00 / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
50.8mm
For separation, fractioning, particle size determination
203.2 mm
5mm
ISO 3310/1
Shaker AS 300
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers