Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes
Brand:  RETSCH 60.149.000900
Additional Details : Weight : 0.35000kg
 View more versions of this product
Product Code. 10360952
  • £106.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
50.8mm
For separation, fractioning, particle size determination
203.2 mm
900μm
ISO 3310/1
Shaker AS 300