missing translation for 'onlineSavingsMsg'
Learn More

Retsch™ 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes

Brand:  Retsch™ 60.140.000300

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11924472

  • £87.00 / Each


Estimated Shipment
27-02-2018
Add to basket

Specifications

Specifications

For separation, fractioning, particle size determination
Stainless Steel
300μm
ISO 3310/1
Shaker AS 300
Stainless Steel
1Each
203.2mm
Test Sieve
Stainless Steel
25.4mm
Videos