Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Pore Sizes: 10mm and Larger
£151.00 - £157.00
Specifications
Diameter (Metric) Outer | 203.2 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11994682
|
RETSCH
60.141.100000 |
100mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11934672
|
RETSCH
60.141.012500 |
12.50mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11984682
|
RETSCH
60.141.106000 |
106mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11924672
|
RETSCH
60.141.011200 |
11.20mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11904692
|
RETSCH
60.141.125000 |
125mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11944672
|
RETSCH
60.141.013200 |
13.20mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11964672
|
RETSCH
60.141.019000 |
19mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11954672
|
RETSCH
60.141.016000 |
16mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11984672
|
RETSCH
60.141.026500 |
26.50mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11974672
|
RETSCH
60.141.022400 |
22.40mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11994672
|
RETSCH
60.141.025000 |
25mm |
£151.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11904682
|
RETSCH
60.141.031500 |
31.50mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11914682
|
RETSCH
60.141.037500 |
37.50mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11924682
|
RETSCH
60.141.045000 |
45mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11944682
|
RETSCH
60.141.053000 |
53mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11934682
|
RETSCH
60.141.050000 |
50mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11954682
|
RETSCH
60.141.063000 |
63mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11964682
|
RETSCH
60.141.075000 |
75mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11974682
|
RETSCH
60.141.090000 |
90mm |
£157.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Specifications
203.2 mm | |
Stainless Steel | |
1Each | |
ASTM E 11 | |
Shaker AS 300 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
25.4mm |