missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ 200mm Diameter Stainless Steel Frame and Wire Mesh Test Sieves

Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Frame Test Sieves can be supplied with aperture sizes ranging from 125mm down to 20 microns in full and half height versions.

Brand:  Endecotts™ 200SIW.600 600UM

Code : 03

Additional Details : Weight : 0.41736kg

 View more versions of this product

Product Code. 12971985

  • £96.00 / Each


In Stock 1
Add to basket

Description

Description

Stainless steel mesh mounted on stainless steel frame, 200mm diameter. All test sieves are supplied with a Certificate of Conformity. Other sizes available on request.
Specifications

Specifications

For all types of laboratory sampling and particle size analysis
Stainless Steel
600μm
ISO 3310-1 & 2 requirements, ASTM, BSI
50mm
1Each
200mm
Test Sieve
Stainless Steel
Stainless Steel
Videos
Documentation View accessory products