missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ 200mm Diameter Brass Frame with Stainless Steel Wire Mesh Test Sieves

Use these test sieves for all types of laboratory sampling and particle size analysis. Endecotts™ Brass Frame Test Sieves can be supplied with aperture sizes ranging from 125mm down to 20 microns in full and half height versions.

Brand:  Endecotts™ 200BIW1.18

Code : 03

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12991955

  • £105.00 / Each


Estimated Shipment
23-02-2018
Add to basket

Description

Description

Brass frame sieves fitted with stainless steel woven wire mesh. All test sieves are supplied with a Certificate of Conformity. 200mm diameter. Other mesh sizes available on request.
Specifications

Specifications

For all types of laboratory sampling and particle size analysis
Brass
1.18mm
ISO 3310-1 & 2 requirements, ASTM, BSI
Stainless Steel
1Each
200mm
Test Sieve
Brass
Videos
Documentation View accessory products