missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S16x50/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S16x50/4MI Swing Out Rotors
£276.00
Specifications
For Use With (Equipment) | R-S16x50/4MI Swing Out Rotors |
---|---|
No. per Pack | 2 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15866652
|
Ohaus™
30314584 |
14 x 5 to 7 mL |
£276.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15876652
|
Ohaus™
30314585 |
12 x 4 to 10 mL |
£276.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856652
|
Ohaus™
30314583 |
10 x 15 mL |
£276.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15846652
|
Ohaus™
30304375 |
4 x 50 mL |
£276.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S16x50/4MI Swing Out Rotors
Specifications
R-S16x50/4MI Swing Out Rotors | |
2 |